Assay Design



Gene Transcripts

Gene Symbol
pde-6
Entrez Gene ID
190808
Gene Name
protein-coding
Gene Aliases
CELE_Y95B8A.10
Chromosome Location
Chr.I: 872755 - 882713 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129083.4 NP_001122555.1 7 - 8 908 81
NM_001306261.3 NP_001293190.1 1 - 2 86 81
NM_001383315.1 7 - 8 908 81

View Full Product Details