Assay Design



Gene Transcripts

Gene Symbol
pde-6
Entrez Gene ID
190808
Gene Name
protein-coding
Gene Aliases
CELE_Y95B8A.10
Chromosome Location
Chr.I: 872755 - 882713 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306261.3 NP_001293190.1 2 - 3 1217 66
NM_001383315.1 8 - 9 2039 66

View Full Product Details