Assay Design
Gene Transcripts
- Gene Symbol
- vps-4
- Entrez Gene ID
-
189590
- Gene Name
- related to yeast Vacuolar Protein Sorting factor
- Gene Aliases
-
CELE_Y34D9A.10
- Chromosome Location
-
Chr.I: 1053894 - 1066905 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058415.8 |
NP_490816.4 |
7 - 8 |
1073 |
75 |
View Full Product Details