Assay Design



Gene Transcripts

Gene Symbol
gsa-1
Entrez Gene ID
171687
Gene Name
protein-coding
Gene Aliases
CELE_R06A10.2
Chromosome Location
Chr.I: 1068484 - 1075036 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380927.1 NP_001367988.1 1 - 2 608 60

View Full Product Details