Assay Design
Gene Transcripts
- Gene Symbol
- emr-1
- Entrez Gene ID
-
171752
- Gene Name
- Emerin homolog 1
- Gene Aliases
-
CELE_M01D7.6
- Chromosome Location
-
Chr.I: 1842445 - 1843227 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058506.6 |
NP_490907.1 |
2 - 3 |
403 |
67 |
View Full Product Details