Assay Design
Gene Transcripts
- Gene Symbol
- tub-2
- Entrez Gene ID
-
171758
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_Y71G12A.3
- Chromosome Location
-
Chr.I: 1874008 - 1884196 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058512.5 |
NP_490913.2 |
1 - 2 |
1291 |
69 |
View Full Product Details