Assay Design
Gene Transcripts
- Gene Symbol
- scrt-1
- Entrez Gene ID
-
183848
- Gene Name
- SCRaTch (zinc finger transcriptional repressor) homolog
- Gene Aliases
-
CELE_C55C2.1
- Chromosome Location
-
Chr.I: 2568860 - 2571059 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058600.3 |
NP_491001.2 |
4 - 5 |
452 |
68 |
View Full Product Details