Assay Design



Gene Transcripts

Gene Symbol
aat-5
Entrez Gene ID
171814
Gene Name
Amino Acid Transporter
Gene Aliases
CELE_C55C2.5
Chromosome Location
Chr.I: 2571740 - 2576020 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058601.6 NP_491002.2 9 - 10 1579 78
NR_131359.1 8 - 9 1622 78
NR_131780.1 9 - 10 1557 78

View Full Product Details