Assay Design



Gene Transcripts

Gene Symbol
fbp-1
Entrez Gene ID
266822
Gene Name
FBPase domain-containing protein
Gene Aliases
CELE_K07A3.1
Chromosome Location
Chr.I: 2580935 - 2587110 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058603.7 NP_491004.1 4 - 5 1066 83
GenBank mRNA AJ271466.1 - 3 - 4 940 83

View Full Product Details