Assay Design
Gene Transcripts
- Gene Symbol
- spg-7
- Entrez Gene ID
-
171915
- Gene Name
- AFG3-like protein spg-7
- Gene Aliases
-
CELE_Y47G6A.10
- Chromosome Location
-
Chr.I: 3467132 - 3478517 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058764.5 |
NP_491165.2 |
6 - 7 |
1830 |
83 |
View Full Product Details