Assay Design
Gene Transcripts
- Gene Symbol
- smp-2
- Entrez Gene ID
-
183905
- Gene Name
- Sema domain-containing protein
- Gene Aliases
-
CELE_D1037.2
- Chromosome Location
-
Chr.I: 3666487 - 3671700 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058796.5 |
NP_491197.3 |
3 - 4 |
446 |
62 |
View Full Product Details