Assay Design
Gene Transcripts
- Gene Symbol
- glh-4
- Entrez Gene ID
-
171941
- Gene Name
- ATP-dependent RNA helicase glh-4
- Gene Aliases
-
CELE_T12F5.3
- Chromosome Location
-
Chr.I: 3726011 - 3731265 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058806.5 |
NP_491207.3 |
4 - 5 |
1740 |
62 |
View Full Product Details