Assay Design
Gene Transcripts
- Gene Symbol
- npp-11
- Entrez Gene ID
-
171954
- Gene Name
- Nuclear Pore complex Protein
- Gene Aliases
-
CELE_F53F10.5
- Chromosome Location
-
Chr.I: 3818074 - 3821508 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058831.7 |
NP_491232.1 |
5 - 6 |
2351 |
74 |
View Full Product Details