Assay Design
Gene Transcripts
- Gene Symbol
- lrp-2
- Entrez Gene ID
-
171981
- Gene Name
- Low-density lipoprotein RecePtor related
- Gene Aliases
-
CELE_T21E3.3
- Chromosome Location
-
Chr.I: 3935272 - 3953991 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058869.5 |
NP_491270.2 |
9 - 10 |
7478 |
123 |
View Full Product Details