Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001026325.2 | NP_001021496.1 | 14 - 15 | 3805 | 105 |
NM_001026326.4 | NP_001021497.1 | 14 - 15 | 3693 | 105 | |
NM_001381834.2 | NP_001367474.1 | 14 - 15 | 3693 | 105 | |
GenBank mRNA | AF048834.1 | - | 14 - 15 | 3805 | 105 |
AF048835.1 | - | 14 - 15 | 3805 | 105 |