Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001026325.2 | NP_001021496.1 | 8 - 9 | 1574 | 84 |
NM_001026326.4 | NP_001021497.1 | 8 - 9 | 1462 | 84 | |
NM_001381834.2 | NP_001367474.1 | 8 - 9 | 1462 | 84 | |
GenBank mRNA | AF048834.1 | - | 8 - 9 | 1574 | 84 |
AF048835.1 | - | 8 - 9 | 1574 | 84 |