Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001026325.2 | NP_001021496.1 | 21 - 22 | 5216 | 65 |
NM_001026327.4 | NP_001021498.1 | 5 - 6 | 750 | 65 | |
NM_001026328.3 | NP_001021499.1 | 2 - 3 | 313 | 65 | |
NM_001026329.6 | NP_001021500.1 | 3 - 4 | 416 | 65 | |
NM_001262583.3 | NP_001249512.1 | 3 - 4 | 409 | 65 | |
NM_001262584.2 | NP_001249513.1 | 5 - 6 | 797 | 65 | |
NM_001375256.3 | NP_001361940.1 | 5 - 6 | 544 | 65 | |
NM_001381834.2 | NP_001367474.1 | 21 - 22 | 5104 | 65 | |
GenBank mRNA | AF048834.1 | - | 21 - 22 | 5216 | 65 |