Assay Design



Gene Transcripts

Gene Symbol
inx-17
Entrez Gene ID
172006
Gene Name
protein-coding
Gene Aliases
CELE_R12E2.4
Chromosome Location
Chr.I: 4165075 - 4167229 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026421.4 NP_001021592.1 3 - 4 577 58
NM_058914.4 NP_491315.1 3 - 4 517 58
GenBank mRNA AF303262.1 - 3 - 4 517 58

View Full Product Details