Assay Design
Gene Transcripts
- Gene Symbol
- lpd-5
- Entrez Gene ID
-
172037
- Gene Name
- LiPid Depleted
- Gene Aliases
-
CELE_ZK973.10
- Chromosome Location
-
Chr.I: 4370441 - 4375343 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001382913.1 |
|
3 - 4 |
516 |
103 |
View Full Product Details