Assay Design



Gene Transcripts

Gene Symbol
dhc-1
Entrez Gene ID
172041
Gene Name
protein-coding
Gene Aliases
CELE_T21E12.4
Chromosome Location
Chr.I: 4386777 - 4401907 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330925.3 NP_001317773.1 3 - 4 2515 73
NM_058962.6 NP_491363.1 6 - 7 5324 73

View Full Product Details