Assay Design
Gene Transcripts
- Gene Symbol
- nrd-1
- Entrez Gene ID
-
172064
- Gene Name
- NRD (yeast Nuclear pre-mRNA Down-regulation) homolog
- Gene Aliases
-
CELE_D1007.7
- Chromosome Location
-
Chr.I: 4597912 - 4602666 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059002.6 |
NP_491403.1 |
7 - 8 |
2720 |
100 |
View Full Product Details