Assay Design
Gene Transcripts
- Gene Symbol
- acbp-1
- Entrez Gene ID
-
172071
- Gene Name
- Acyl-CoA-binding protein homolog 1
- Gene Aliases
-
CELE_C44E4.6
- Chromosome Location
-
Chr.I: 4635743 - 4637328 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059011.7 |
NP_491412.1 |
3 - 3 |
286 |
83 |
View Full Product Details