Assay Design
Gene Transcripts
- Gene Symbol
- ugt-25
- Entrez Gene ID
-
172087
- Gene Name
- UDP-glucuronosyltransferase
- Gene Aliases
-
CELE_C10H11.3
- Chromosome Location
-
Chr.I: 4731603 - 4735276 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059037.5 |
NP_491438.1 |
4 - 5 |
977 |
125 |
View Full Product Details