Assay Design
Gene Transcripts
- Gene Symbol
- kca-1
- Entrez Gene ID
-
172090
- Gene Name
- Kinesin Cargo Adaptor
- Gene Aliases
-
CELE_C10H11.10
- Chromosome Location
-
Chr.I: 4759820 - 4761321 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059042.6 |
NP_491443.3 |
1 - 2 |
543 |
61 |
GenBank mRNA |
DQ178243.1 |
- |
1 - 2 |
545 |
61 |
View Full Product Details