Assay Design



Gene Transcripts

Gene Symbol
gnrr-1
Entrez Gene ID
186230
Gene Name
protein-coding
Gene Aliases
CELE_F54D7.3
Chromosome Location
Chr.I: 4790290 - 4804393 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001262791.3 NP_001249720.1 2 - 3 396 108
NM_001262792.3 NP_001249721.1 6 - 7 890 108

View Full Product Details