Assay Design



Gene Transcripts

Gene Symbol
him-1
Entrez Gene ID
172116
Gene Name
protein-coding
Gene Aliases
CELE_F28B3.7
Chromosome Location
Chr.I: 4951570 - 4957667 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047193.5 NP_001040658.2 9 - 10 3243 90

View Full Product Details