Assay Design
Gene Transcripts
- Gene Symbol
- slx-1
- Entrez Gene ID
-
172157
- Gene Name
- Structure-specific endonuclease subunit SLX1 homolog
- Gene Aliases
-
CELE_F56A3.2
- Chromosome Location
-
Chr.I: 5182736 - 5186240 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059140.6 |
NP_491541.1 |
1 - 2 |
115 |
151 |
View Full Product Details