Assay Design



Gene Transcripts

Gene Symbol
erm-1
Entrez Gene ID
172174
Gene Name
protein-coding
Gene Aliases
CELE_C01G8.5
Chromosome Location
Chr.I: 5284026 - 5291604 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059158.7 NP_491559.1 9 - 10 1522 67
NM_059159.4 NP_491560.1 9 - 10 1488 67
GenBank mRNA AB107268.1 - 9 - 10 1544 67
AB107269.1 - 9 - 10 1510 67
AY643538.1 - 9 - 10 1512 67

View Full Product Details