Assay Design
Gene Transcripts
- Gene Symbol
- ppk-1
- Entrez Gene ID
-
172184
- Gene Name
- PIPK domain-containing protein
- Gene Aliases
-
CELE_F55A12.3
- Chromosome Location
-
Chr.I: 5358522 - 5392671 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059175.6 |
NP_491576.2 |
7 - 8 |
1205 |
101 |
View Full Product Details