Assay Design
Gene Transcripts
- Gene Symbol
- rpl-19
- Entrez Gene ID
-
172201
- Gene Name
- 60S ribosomal protein L19
- Gene Aliases
-
CELE_C09D4.5
- Chromosome Location
-
Chr.I: 5483132 - 5486354 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059207.8 |
NP_491608.1 |
4 - 5 |
575 |
118 |
View Full Product Details