Assay Design
Gene Transcripts
- Gene Symbol
- ptr-10
- Entrez Gene ID
-
172229
- Gene Name
- SSD domain-containing protein
- Gene Aliases
-
CELE_F55F8.1
- Chromosome Location
-
Chr.I: 5662284 - 5667160 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383129.1 |
|
11 - 12 |
1904 |
94 |
View Full Product Details