Assay Design
Gene Transcripts
- Gene Symbol
- unc-40
- Entrez Gene ID
-
172233
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_T19B4.7
- Chromosome Location
-
Chr.I: 5679837 - 5691503 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059263.10 |
NP_491664.1 |
6 - 7 |
772 |
84 |
GenBank mRNA |
U70618.1 |
- |
6 - 7 |
714 |
84 |
View Full Product Details