Assay Design
Gene Transcripts
- Gene Symbol
- npp-7
- Entrez Gene ID
-
172234
- Gene Name
- Nuclear Pore complex Protein
- Gene Aliases
-
CELE_T19B4.2
- Chromosome Location
-
Chr.I: 5692264 - 5697506 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383135.1 |
|
1 - 2 |
200 |
97 |
View Full Product Details