Assay Design
Gene Transcripts
- Gene Symbol
- lin-35
- Entrez Gene ID
-
172249
- Gene Name
- Retinoblastoma-like protein homolog lin-35
- Gene Aliases
-
CELE_C32F10.2
- Chromosome Location
-
Chr.I: 5807709 - 5815637 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059285.4 |
NP_491686.1 |
1 - 2 |
79 |
62 |
GenBank mRNA |
AF116529.1 |
- |
1 - 2 |
101 |
62 |
View Full Product Details