Assay Design
Gene Transcripts
- Gene Symbol
- tyr-4
- Entrez Gene ID
-
172263
- Gene Name
- TYRosinase
- Gene Aliases
-
CELE_C34G6.2
- Chromosome Location
-
Chr.I: 5898303 - 5902118 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059308.8 |
NP_491709.1 |
2 - 3 |
286 |
73 |
View Full Product Details