Assay Design
Gene Transcripts
- Gene Symbol
- ntl-4
- Entrez Gene ID
-
177580
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_C49H3.5
- Chromosome Location
-
Chr.IV: 7910575 - 7915189 on Build WBcel235
- Gene Symbol
- C49H3.4
- Entrez Gene ID
-
183627
- Gene Name
- Sm domain-containing protein
- Gene Aliases
-
CELE_C49H3.4
- Chromosome Location
-
Chr.IV: 7910604 - 7916711 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383131.1 |
|
9 - 10 |
1150 |
113 |
View Full Product Details