Assay Design
Gene Transcripts
- Gene Symbol
- pas-5
- Entrez Gene ID
-
172942
- Gene Name
- Proteasome subunit alpha type-5
- Gene Aliases
-
CELE_F25H2.9
- Chromosome Location
-
Chr.I: 10566350 - 10567300 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060364.7 |
NP_492765.1 |
1 - 2 |
229 |
71 |
View Full Product Details