Assay Design
Gene Transcripts
- Gene Symbol
- lam-3
- Entrez Gene ID
-
172952
- Gene Name
- LAMinin related. See also lmb-
- Gene Aliases
-
CELE_T22A3.8
- Chromosome Location
-
Chr.I: 10608466 - 10621435 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060374.5 |
NP_492775.2 |
2 - 3 |
863 |
143 |
GenBank mRNA |
AF074902.1 |
- |
2 - 3 |
863 |
143 |
View Full Product Details