Assay Design



Gene Transcripts

Gene Symbol
pst-1
Entrez Gene ID
178980
Gene Name
protein-coding
Gene Aliases
CELE_M03F8.2
Chromosome Location
Chr.V: 5943467 - 5951417 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373028.2 NP_001360551.1 2 - 2 56 67
NM_001383312.1 1 - 2 51 67
NM_171463.5 NP_741545.2 1 - 2 49 67

View Full Product Details