Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001028889.4 | NP_001024060.1 | 4 - 5 | 841 | 74 |
NM_001373028.2 | NP_001360551.1 | 4 - 5 | 818 | 74 | |
NM_171463.5 | NP_741545.2 | 4 - 5 | 811 | 74 | |
NM_171464.5 | NP_741546.3 | 4 - 5 | 1173 | 74 |