Assay Design



Gene Transcripts

Gene Symbol
pst-1
Entrez Gene ID
178980
Gene Name
protein-coding
Gene Aliases
CELE_M03F8.2
Chromosome Location
Chr.V: 5943467 - 5951417 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028889.4 NP_001024060.1 5 - 6 950 137
NM_001373028.2 NP_001360551.1 5 - 6 927 137
NM_001383312.1 5 - 6 918 137
NM_171463.5 NP_741545.2 5 - 6 920 137
NM_171464.5 NP_741546.3 5 - 6 1282 137

View Full Product Details