Assay Design



Gene Transcripts

Gene Symbol
atg-18
Entrez Gene ID
179246
Gene Name
protein-coding
Gene Aliases
CELE_F41E6.13
Chromosome Location
Chr.V: 8629174 - 8631327 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171491.5 NP_741576.1 4 - 5 817 68
NM_171492.6 NP_741577.1 4 - 5 819 68

View Full Product Details