Assay Design



Gene Transcripts

Gene Symbol
xbx-6
Entrez Gene ID
179361
Gene Name
protein-coding
Gene Aliases
CELE_F40F9.1
Chromosome Location
Chr.V: 9703055 - 9707835 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373079.3 NP_001359646.1 4 - 4 404 82
NM_171511.7 NP_741597.1 4 - 5 406 82
NR_069217.1 4 - 5 409 82

View Full Product Details