Assay Design



Gene Transcripts

Gene Symbol
egl-9
Entrez Gene ID
179461
Gene Name
protein-coding
Gene Aliases
CELE_F22E12.4
Chromosome Location
Chr.V: 10468517 - 10476915 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028663.6 NP_001023834.1 1 - 2 86 93
NM_001373098.1 NP_001359645.1 1 - 2 86 93
NM_171533.6 NP_741621.1 1 - 2 86 93
GenBank mRNA AF178536.1 - 1 - 2 88 93

View Full Product Details