Assay Design



Gene Transcripts

Gene Symbol
dmsr-1
Entrez Gene ID
186437
Gene Name
protein-coding
Gene Aliases
CELE_F57B7.1
Chromosome Location
Chr.V: 11438764 - 11443450 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171541.6 NP_741629.1 1 - 2 108 124
NM_171542.7 NP_741630.1 1 - 2 114 124

View Full Product Details