Assay Design
Gene Transcripts
- Gene Symbol
- dmsr-1
- Entrez Gene ID
-
186437
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F57B7.1
- Chromosome Location
-
Chr.V: 11438764 - 11443450 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_171542.7 |
NP_741630.1 |
5 - 6 |
1339 |
129 |
View Full Product Details