Assay Design



Gene Transcripts

Gene Symbol
max-1
Entrez Gene ID
179794
Gene Name
protein-coding
Gene Aliases
CELE_C34B4.1
Chromosome Location
Chr.V: 13098903 - 13120220 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380794.1 NP_001367137.1 7 - 8 1804 99
NM_001383435.1 8 - 9 1946 99
GenBank mRNA AY084080.1 - 7 - 8 1804 99

View Full Product Details