Assay Design



Gene Transcripts

Gene Symbol
hot-1
Entrez Gene ID
179966
Gene Name
protein-coding
Gene Aliases
CELE_F43D2.4
Chromosome Location
Chr.V: 14621135 - 14629472 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136425.3 NP_001129897.1 2 - 3 191 73
NM_171552.4 NP_741640.1 2 - 3 314 73
GenBank mRNA AF324053.1 - 2 - 3 308 73

View Full Product Details