Assay Design
Gene Transcripts
- Gene Symbol
- twk-46
- Entrez Gene ID
-
180252
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_Y39B6A.19
- Chromosome Location
-
Chr.V: 19060173 - 19067034 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_171588.6 |
NP_741678.1 |
2 - 3 |
249 |
76 |
View Full Product Details