Assay Design



Gene Transcripts

Gene Symbol
twk-46
Entrez Gene ID
180252
Gene Name
protein-coding
Gene Aliases
CELE_Y39B6A.19
Chromosome Location
Chr.V: 19060173 - 19067034 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136444.3 NP_001129916.1 2 - 3 445 78
NM_171588.6 NP_741678.1 3 - 4 455 78

View Full Product Details